iFluor® 800, Succinimidyl Ester
Dodavatel: AAT BIOQUEST
1379
71379
71525
71575
AATB1379EA
11800
CZK
AATB1379
AATB71379
AATB71525
AATB71575
iFluor® 800, Succinimidyl Ester
iFluor®
In vivo fluorescence imaging uses a sensitive camera to detect fluorescence emission from fluorophores in whole-body living small animals.
- Bright fluorescence
- High photostability
- Low quenching on proteins
- Useful for in-vivo imaging
Upozornení: For research use only.