Tisk…

5(6)-Carboxyfluorescein ≥95%

Dodavatel: AAT BIOQUEST
Varování

Synonyms: 5(6)-FAM, 3′,6′-Dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthene]-ar-carboxylic acid

100 101 102
AATB100EA 4060 CZK
AATB100 AATB101 AATB102
5(6)-Carboxyfluorescein ≥95%
5(6)-Carboxyfluorescein

Carboxyfluorescein (commonly called FAM) and its amine-reactive succinimidyl esters are favored over FITC in bioconjugations.

Vzorec: C₄₂H₂₄O₁₄
MW: 752,63 g/mol
MDL Number: MFCD00151081
CAS číslo: 72088-94-9
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c266e82127bb9214ac06e1da48806d25cdeaa4a&vid=16b52fecf5798aaa1efb60ac1f30d13a87cdad70&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR