Tisk…
ESD T-Shirts, AS22
Dodavatel: Portwest
AS22NARS AS22NARL AS22NARM AS22NARXL AS22NARXXL
PRTWAS22NARSEA 956 CZK
PRTWAS22NARS PRTWAS22NARL PRTWAS22NARM PRTWAS22NARXL PRTWAS22NARXXL
ESD T-Shirts, AS22
Pracovní oděvy
Protective long sleeve t-shirt is light and comfortable and it is made up of 96% Cotton, 4% Carbon fiber. Ideal for environments where electrostatic resistance is required. Carbon fibre woven through the fabric provides superb antistatic protection.
  • Suitable for use in ESD environments
  • Designed with a comfort fit
  • Lightweight and comfortable
  • Ribbed cuffs for added comfort
  • Ribbed collar
  • Anti-static
Fabrics used in the ESD range are strong and durable. Carbon fibre woven through the fabric provides superb anti static protection.
Certifikace: Tested to EN 61340-5-1, EN 1149-5. UKCA marked.