Tisk…
Vícepolohové ohřívače míchacího bloku, Drybath 15 - 100 a 15 - 250
Dodavatel: 2MAG
63250 63250_CH 63250_UK 63100_UK 63100_CH 63100
460-0189EA 382670 CZK
460-0189 460-0190 460-0191 460-0188 460-0187 460-0186
Vícepolohové ohřívače míchacího bloku, Drybath 15 - 100 a 15 - 250
Termostaty blokové Termostaty blokové/ohřívací
Ideal for viscosity measurements, testing and quality control. The units have a clear digital display for settings of stirrer speed, stirrer power and temperature of the integrated heater. The SoftStart procedure ensures reliable centering and safe acceleration of the stirring bar. The heating block is made of aluminium alloy, PTFE coated for chemical resistance and easy cleaning.
  • Maintenance- and wear-free inductive drive
  • Speed range of 100 to 2000 rpm
  • Stirrer has ten step power settings
  • 100% synchronised, jerk-free stirring
  • Independent over-temperature protection inside the heating plate
15 - 100
Suitable for 100 ml flasks and 150 ml beaker glasses
15 holes with Ø 57,5 mm and 48 mm depth
65 mm between stirring points
15 - 250
Suitable for 250 ml flasks and 400 ml beaker glasses
15 holes with Ø 71,5 mm and 78 mm depth
84 mm between stirring points
Informace o dodávce: Supplied with 2mag heatMIXcontrol control unit and adjustable over-temperature limiter for the additional protection of the heating controller.