Tisk…
SteriWare Graduated Beakers
Dodavatel: Sampling Systems
8008P-250S 8008P-50S 8008P-2000 8008P-2000S 8008P-250 8008P-100 8008P-50 8008P-25S 8008P-100S 8008P-25
SAMP8008P-250SEA 9100 CZK
SAMP8008P-250S SAMP8008P-50S SAMP8008P-2000 SAMP8008P-2000S SAMP8008P-250 SAMP8008P-100 SAMP8008P-50 SAMP8008P-25S SAMP8008P-100S SAMP8008P-25
SteriWare Graduated Beakers
Kádinky
The SteriWare range of greduated beakers are ideal for use in the production of drugs, chemicals and cosmetics.
  • Made from food grade, FDA and EC compliant PP
  • Individually bagged
  • Choice of sterile and non sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom
The range of individually bagged graduated beakers from sampling systems are crucial in the production of drugs, chemicals and cosmetics. As they are indivicually bagged they prevent cross contamination and they have graduation marks and a pouring spout to ensure the convnenient handling of liquids.