Tisk…

Anti-Immunotag™ HXK I Mouse Monoclonal Antibody (Alexa Fluor® 555)

Dodavatel: G-Biosciences
ITM0349-50U555 ITM0349100U555
GENOITM0349-50U555EA 25650 CZK
GENOITM0349-50U555 GENOITM0349100U555
Anti-Immunotag™ HXK I Mouse Monoclonal Antibody (Alexa Fluor® 555)
Protilátky
Hexokinase 1(HK1) Homo sapiens Hexokinases phosphorylate glucose to produce glucose-6-phosphate, the first step in most glucose metabolism pathways. This gene encodes a ubiquitous form of hexokinase which localises to the outer membrane of mitochondria. Mutations in this gene have been associated with hemolytic anemia due to hexokinase deficiency. Alternative splicing of this gene results in several transcript variants which encode different isoforms, some of which are tissue-specific.

  • Application: WB, IHC-p, IF, FCM, ELISA
  • Recommended dilution: Western blot: 1/500 - 1/2000, Immunohistochemistry: 1/200 - 1/1000, Immunofluorescence: 1/200 - 1/1000, Flow cytometry: 1/200 - 1/400, ELISA: 1/10000

Upozornení: For research use only. Not for diagnostic or therapeutic procedures.

Type:
Antigen: Hexokinase-1
Clonality: Monoclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR