Tisk…
Bizflame Industry Two-Tone Jackets
Dodavatel: Portwest
FR08RBRL FR08RBRM FR08RBRS FR08RBRXL FR08RBRXXL FR08RBRXXXL
PRTWFR08RBRLEA 3900 CZK
PRTWFR08RBRL PRTWFR08RBRM PRTWFR08RBRS PRTWFR08RBRXL PRTWFR08RBRXXL PRTWFR08RBRXXXL
Bizflame Industry Two-Tone Jackets
Pracovní oděvy
Designed for working in hazardous conditions both off-shore or on-site, this jacket combines style with performance. The special blend of FR polyester in the fabric provides unbeatable durability in the toughest conditions.
  • Hook and loop cuffs for a secure fit
  • Adjustable waist for a perfect fit
  • Non magnetic; nickel- and ferrous-free
  • 40+ UPF rated fabric to block 98% of UV rays
  • Sew-on flame-resistant industrial wash tape
  • Radio loops for easy clipping of a radio
  • Five pockets for ample storage
Jackets provide protection against radiant, convective, and contact heat, as well as certified protection against molten metal splash. An external standards label enables quick identification of protection levels.
Industrial laundry wash at 75 °C. Tunnel dry at 155 °C.
Certifikace: EN ISO 11612 A1+A2, B1, C1, E2, F1. EN ISO 11611 Class 1 A1+A2. EN 1149 -5. IEC 61482-2 IEC 61482-1-1 Elim 11 cal/cm². IEC 61482-2 IEC 61482-1-2 APC 1. EN 13034 Type PB [6]. ASTM F1959/F1959M-12 ATPV 16 cal/cm² (HAF 83.7%).