Tisk…
Erlenmeyer Flasks, Narrow Neck
Dodavatel: GLASSCO
231.202.01 231.202.02 231.202.05 231.202.06 231.202.07
GLAS231.202.01EA 53.6 CZK
GLAS231.202.01 GLAS231.202.02 GLAS231.202.05 GLAS231.202.06 GLAS231.202.07
Erlenmeyer Flasks, Narrow Neck
Baňky Baňky dle Erlenmeyera
Clear, conical, graduated and narrow mouth Erlenmeyer flask, made from ASTM E-438 Type 1, Borosilicate 3.3 heat resistance glass.
  • All the flasks are printed in white enamel
  • Has reinforced thick rim to reduce chipping and breakage
Certifikace: Complies with ISO 1773.