Tisk…
LONDON Stretch Trousers
Dodavatel: ALSICO
1035655-BE38 1035655-BE40 1035655-BE42 1035655-BE44 1035655-BE46 1035655-BE48 1035655-BE50 1035655-BE52 1035655-BE54 1035655-BE56 1035655-BE58 1035655-BE60 1035655-BE62 1035655-BE64 1035865-BE38 1035865-BE40 1035865-BE42 1035865-BE44 1035865-BE46 1035865-BE48 1035865-BE50 1035865-BE52 1035865-BE54 1035865-BE56 1035865-BE58 1035865-BE60 1035865-BE62 1035865-BE64
ALSW1035655-BE38EA 0 CZK
ALSW1035655-BE38 ALSW1035655-BE40 ALSW1035655-BE42 ALSW1035655-BE44 ALSW1035655-BE46 ALSW1035655-BE48 ALSW1035655-BE50 ALSW1035655-BE52 ALSW1035655-BE54 ALSW1035655-BE56 ALSW1035655-BE58 ALSW1035655-BE60 ALSW1035655-BE62 ALSW1035655-BE64 ALSW1035865-BE38 ALSW1035865-BE40 ALSW1035865-BE42 ALSW1035865-BE44 ALSW1035865-BE46 ALSW1035865-BE48 ALSW1035865-BE50 ALSW1035865-BE52 ALSW1035865-BE54 ALSW1035865-BE56 ALSW1035865-BE58 ALSW1035865-BE60 ALSW1035865-BE62 ALSW1035865-BE64
LONDON Stretch Trousers
Pracovní oděvy
These trousers for men are a modern take on the traditional chino, offering a professional look without compromising comfort. The fabric is powered by Alsiflex technology, which provides a unique combination of stretch, sustainability, and performance.
  • Waistband with elastic in the side
  • Front closure with jeans button and zipper
  • 2 large side pockets
  • 1 back pocket
  • Inserted phone pocket on back leg
  • Extendable trouser legs
Industrial launderable.
Certifikace: Oeko-Tex® standard 100.
Objednat
Produktová specifikace
Environmentally Preferable
Aplikace oblečení Industrial,Mechanical protection
Risk area
Rod Pro muže
Sleeve or leg length Long
Typ uzávěru Se zipem
Tkanina Alsiflex (79% polyester, 21% cotton)
Hmotnost 265 g/m²