N-Biotinyl-ethylenediamine trifluoroacetate salt ≥96.5% (by HPLC), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
08599-100MG
08599-500MG
SIAL08599-100MGEA
12200
CZK
SIAL08599-100MG
SIAL08599-500MG
N-Biotinyl-ethylenediamine trifluoroacetate salt ≥96.5% (by HPLC), Sigma-Aldrich®
N-Biotinyl-ethylenediamine trifluoroacetate salt
Nucleophilic biotinylation reagent for the conjugation of various substrates and their identification and quantification using biotin-binding fluorescent labels
Vzorec:
C₁₂H₂₂N₄O₂S·C₂HF₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11045268 CAS číslo: 1217450-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...