Tisk…
PermaSleeve® Commercial Grade Wire Markers for M610, M611, BMP61, M710 and BMP71
Dodavatel: Brady
174435 174423 174458 174417 174441 174463
BRDY174435EA 10030 CZK
BRDY174435 BRDY174423 BRDY174458 BRDY174417 BRDY174441 BRDY174463
PermaSleeve® Commercial Grade Wire Markers for M610, M611, BMP61, M710 and BMP71
Štítky
PermaSleeve® commercial grade wire markers provide business-class performance at a commercial price, so users can easily complete wire identification projects throughout their facilities.
  • Great for electrical contractors, maintenance crews, industrial manufacturers and more
  • Heat-shrink sleeves provide permanent identification without any extra steps
  • Legible legends in temperatures ranging from −55 to +135 °C (−67 to +275 °F)
  • Matte finish
Applications include electrical labelling and wire and cable labelling.
A ribbon is required to print this material. Use part number M61-R4310 for the M610/BMP61/Brady printer M611/M611 label printers or M7-R4300 for the M710/BMP71 label printers.
Certifikace: UL 224 recognised.