Tisk…

Bole white (kaolin), powder USP

Dodavatel: Spectrum Chemical

Synonyms: China Clay, Hydrated aluminum silicate, Clay

KA105-12KG KA105-2.5KG
SPCMKA105-12KGEA 36460 CZK
SPCMKA105-12KG SPCMKA105-2.5KG
Bole white (kaolin), powder USP
Kaolin

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: Al₂SiO₇·2H₂O
Bod tání: 1760 °C
Hustota: 0,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00062311
CAS číslo: 1332-58-7
EINECS: 310-194-1
REACH: exempt
Merck Index: 13,05300

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR