Tisk…

Cetostearyl alcohol NF

Dodavatel: Spectrum Chemical

C1888-12KG C1888-2.5KG C1888-45KGBL
SPCMC1888-12KGEA 96510 CZK
SPCMC1888-12KG SPCMC1888-2.5KG SPCMC1888-45KGBL
Cetostearyl alcohol NF
Cetostearyl alcohol

Cetostearyl Alcohol, NF is used as a stabiliser and opacifying agent.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Storage Temperature: Ambient
MDL Number: MFCD04113584
CAS číslo: 8005-44-5

Objednat

Výsledky specifikačního testu

Stearyl Alcohol and Cetyl Alcohol 90.0 - 102.0%
Stearyl Alcohol 40.0 % min.
Hydroxyl Value 208 - 228
Acid Value max. 2
Iodine Value max. 4


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR