Tisk…

Castor oil ethoxylated

Dodavatel: Spectrum Chemical

Synonyms: Polyoxyl 35 castor oil, Castor oil polyoxyl 35, PEG 35 castor oil

C2305-1KG C2305-20KG C2305-60KGBL
SPCMC2305-1KGEA 26510 CZK
SPCMC2305-1KG SPCMC2305-20KG SPCMC2305-60KGBL
Castor oil ethoxylated
Castor oil ethoxylated

Castor Oil, Ethoxylated is an oil that is derived from the castor beans that has been polyethoxylated. A colorless oil that has no taste or smell and boils at 313 C, it is used in the production of many products such as lubricants, paints, dyes, waxes, perfumes, pharmaceuticals and plastics.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Hustota: 1,050
Storage Temperature: Ambient
MDL Number: MFCD00130879
CAS číslo: 61791-12-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR