Tisk…
N-Acetyl-L-tyrosine
Dodavatel: Spectrum Chemical
Synonyms: N-Acetyl-L-Tyrosine, (S)-2-Acetamido-3-(4-hydroxyphenyl)propanoic acid, Ac-Tyr-OH
A2017-1KG A2017-25KGBL A2017-5KGBL
SPCMA2017-1KGEA 0 CZK
SPCMA2017-1KG SPCMA2017-25KGBL SPCMA2017-5KGBL
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine
N-Acetyl-L-tyrosine, also known as 4-hydroxyphenylalanine, is used by cells to synthesise proteins.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Vzorec: C₁₁H₁₃NO₄
MW: 223.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037190
CAS číslo: 537-55-3