Tisk…
D-ɑ-Tocopherol succinate ≥96.0% USP
Dodavatel: Spectrum Chemical
Synonyms: Vitamin E Succinate, D-α-Tocopherol Succinate, 4-Oxo-4-(((R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl)oxy)butanoic acid
V1035-1KG V1035-25KGBL
SPCMV1035-1KGEA 54540 CZK
SPCMV1035-1KG SPCMV1035-25KGBL
D-ɑ-Tocopherol succinate ≥96.0% USP
D-alpha-Tocopherol succinate
RRR-Vitamin E Succinate, 1 mg = 0.81 alpha-tocopherol equivalents, USP.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Vzorec: C₃₃H₅₄O₅
MW: 530.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00072055
CAS číslo: 4345-03-3
Objednat
Výsledky specifikačního testu
Assay 96.0 - 102.0 %
Acidity To Pass Test
Identification To Pass Test