Tisk…

Choline dihydrogen citrate ≥98%

Dodavatel: Spectrum Chemical

C1229-12KG C1229-25KGBL C1229-50KGBL
SPCMC1229-12KGEA 112440 CZK
SPCMC1229-12KG SPCMC1229-25KGBL SPCMC1229-50KGBL
Choline dihydrogen citrate ≥98%
Choline dihydrogen citrate

Choline Dihydrogen Citrate is part of a class of quaternary ammonium salts containing the N,N,N-trimethylethanolammonium cation and Dihydrogen Citrate counteranion. Choline is the precursor for the neurotransmitter acetylcholine, which is involved in many functions including memory and muscle control. This item is of known assay and suitable for most chemical uses.


GHS hazard : Exclamation mark

Vzorec: C₁₁H₂₁NO₈
Storage Temperature: Ambient
MDL Number: MFCD00038734
CAS číslo: 77-91-8

Objednat

Výsledky specifikačního testu

Assay 98% min.
Water max.0.25 %
Residue On Ignition max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR