Tisk…
Coconut oil, refined
Dodavatel: Spectrum Chemical
CO110-12KG CO110-2.5KG CO110-200LTBL
SPCMCO110-12KGEA 22460 CZK
SPCMCO110-12KG SPCMCO110-2.5KG SPCMCO110-200LTBL
Coconut oil, refined
Coconut oil
Coconut Oil, Refined is taken from the meat of coconuts that come from the coconut palm. It is considered an edible oil and has several purposes in food, industry, and medicine. It contains high saturated fat.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature: Ambient
MDL Number: MFCD00130817
CAS číslo: 8001-31-8