Tisk…

Carboxymethyl cellulose sodium salt, 10-50 cP (2 % H₂O 25°C) USP, low viscosity

Dodavatel: Spectrum Chemical

Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

CA193-125GM CA193-2.5KG CA193-500GM
SPCMCA193-125GMEA 4880 CZK
SPCMCA193-125GM SPCMCA193-2.5KG SPCMCA193-500GM
Carboxymethyl cellulose sodium salt, 10-50 cP (2 % H₂O 25°C) USP, low viscosity
Carboxymethyl cellulose sodium salt

Carboxymethylcellulose Sodium, Low Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 37.5 cP, USP is often used as a thickener and stabiliser. The USP grade indicates it is a grade suitable for personal care, cosmetic and pharmaceutical applications and were manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Vzorec: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS číslo: 9004-32-4

Objednat

Výsledky specifikačního testu

Actual Viscosity of 2% Aqueous Solution @ 25°C As Reported
Sodium (Dried Basis) 6.5 - 9.5%
pH of a 1 % Solution @ 25°C 6.5 - 8.5
Loss On Drying max. 10.0%
Heavy Metals max. 20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR