Tisk…

Sucralose NF

Dodavatel: Spectrum Chemical

S1416-10KGBL S1416-2.5KGBL S1416-500GM S1416-25KGBL
SPCMS1416-10KGBLEA 0 CZK
SPCMS1416-10KGBL SPCMS1416-2.5KGBL SPCMS1416-500GM SPCMS1416-25KGBL
Sucralose NF
Sucralose

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Vzorec: C₁₂H₁₉Cl₃O₈
Bod tání: 130 °C
Storage Temperature: Ambient
MDL Number: MFCD03648615
CAS číslo: 56038-13-2

Objednat

Výsledky specifikačního testu

APPEARANCE CONFORM
ASSAY (ANHYDROUS BASIS) 98.0 - 102.0 %
SPECIFIC ROTATION +84.0° to +87.5°
WATER DETERMINATION Max. 2.0 %
RESIDUE ON IGNITION Max. 0.7 %
ELEMENTAL IMPURITIES:
CADMIUM (Cd) AS REPORTED
LEAD (Pb) AS REPORTED
ARSENIC (As) AS REPORTED
MERCURY (Hg) AS REPORTED
HYDROLYSIS PRODUCTS Max. 0.1 %
METHANOL Max. 0.1 %
RELATED COMPOUNDS Max. 0.5 %
IDENTIFICATION A (FTIR) SPECTRUM MATCHES REFERENCE
IDENTIFICATION (B) RETENTION TIME CORRESPONDS TO STANDARD
IDENTIFICATION (C) Rf VALUE CORRESPONDS TO STANDARD
CERTIFIED HALAL
RESIDUAL SOLVENTS AS REPORTED


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR