NF-κB Activation Inhibitor III ≥99%
Dodavatel: Cayman Chemical
19083-25
19083-5
CAYM19083-25EA
15200
CZK
CAYM19083-25
CAYM19083-5
NF-κB Activation Inhibitor III ≥99%
NF-κB Activation Inhibitor III
- Inhibitor of TNF-α-induced MMP-9 protein expression
Inhibits TNF-α-induced MMP-9 protein expression (maximal effect at 10 µM) by blocking NF-κB activity in HT1080 human fibrosarcoma cells.
Vzorec:
C₁₀H₅ClN₄O₅S Storage Temperature: Freezer |
MDL Number:
MFCD00834007 CAS číslo: 380623-76-7 |
Výsledky specifikačního testu
Formulation | A white to off-white solid |
Purity | ≥99% |
Stability | ≥ 1 year |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...