Tisk…

CID-2858522 ≥98%

Dodavatel: Cayman Chemical
Nebezpečí

Synonyms: ML-029, 1-[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]-2-[2-[(3-hydroxypropyl)amino]-5,6-dimethyl-1H-benzimidazol-1-yl]-ethanone

14122-1 14122-25 14122-5 14122-10
CAYM14122-1EA 1540 CZK
CAYM14122-1 CAYM14122-25 CAYM14122-5 CAYM14122-10
CID-2858522 ≥98%
CID-2858522

  • A selective inhibitor of NF-κB activation.


A selective inhibitor of PKC-induced NF-κB pathway activation (IC₅₀= 0,07 μM); inhibits IL-8 production in HEK293 cells (IC₅₀ ≤0,1 μM), attenuates production of IL-2 by Jurkat T cells (IC₅₀ ≤5 μM), and prevents proliferation of mouse B-lymphocytes (IC₅₀= ∼2 μM)

Vzorec: C₂₈H₃₉N₃O₃
MW: 465.6 g/mol
Storage Temperature: Freezer
CAS číslo: 758679-97-9
UN: 2811
ADR: 6.1 III

Objednat

Výsledky specifikačního testu

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR