Tisk…
BENTE XXE GTX S3 ESD Safety Shoes, Lace-Up, Low
Dodavatel: ELTEN
720721/40 720721/41 720721/42 720721/43 720721/44 720721/45 720721/46 720721/47 720721/48
ELTN720721/40EA 4050 CZK
ELTN720721/40 ELTN720721/41 ELTN720721/42 ELTN720721/43 ELTN720721/44 ELTN720721/45 ELTN720721/46 ELTN720721/47 ELTN720721/48
BENTE XXE GTX S3 ESD Safety Shoes, Lace-Up, Low
Obuv
Safety shoe, BENTE XXE GTX, lace-up design in blue finish.
  • Hydrophobized textile material
  • GORE-TEX extended comfort footwear
  • Padded tongue
  • Full-length inlay sole ESD
  • Soft fleece ESD insole
  • Non-metallic, penetration-resistant midsole
  • Rubber/EVA sole WELLMAXX ENERGETIC
  • Steel toe cap
For indoor and outdoor use, in areas where exposure to moisture is expected (S2), areas where there is a risk of penetration from pointed and sharp objects (S3), or areas where there is a risk of electrostatic discharge (ESDS/ESD).
Certifikace: EN ISO 20345 S3 HRO/CI/HI/SRC, form A.