Tisk…

3-Bromo-4-(difluoromethyl)iodobenzene ≥95%

Dodavatel: Apollo Scientific

Synonyms: 2-Bromo-1-(difluoromethyl)-4-iodobenzene

PC50181-250MG PC50181-1G
APOSPC50181-250MGEA 8070 CZK
APOSPC50181-250MG APOSPC50181-1G
3-Bromo-4-(difluoromethyl)iodobenzene ≥95%
3-Bromo-4-(difluoromethyl)iodobenzene
Vzorec: C₇H₄BrF₂I
MW: 332,91193 g/mol
MDL Number: MFCD18391110
CAS číslo: 1261569-90-7
UN: 3265

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR