4-Bromo-3,5-difluorobenzyl bromide ≥95%
Dodavatel: Apollo Scientific
PC501916-250MG
PC501916-1G
PC501916-5G
APOSPC501916-250MGEA
5920
CZK
APOSPC501916-250MG
APOSPC501916-1G
APOSPC501916-5G
4-Bromo-3,5-difluorobenzyl bromide ≥95%
4-Bromo-3,5-difluorobenzyl bromide
Vzorec:
C₇H₄Br₂F₂ MW: 285.91146 g/mol |
MDL Number:
MFCD22381221 CAS číslo: 1571115-92-8 UN: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...