2-Bromo-1-(4-bromo-3,5-difluorophenyl)ethanone ≥95%
Dodavatel: Apollo Scientific
PC57413-1G
PC57413-5G
APOSPC57413-1GEA
5560
CZK
APOSPC57413-1G
APOSPC57413-5G
2-Bromo-1-(4-bromo-3,5-difluorophenyl)ethanone ≥95%
2-Bromo-1-(4-bromo-3,5-difluorophenyl)ethanone
Vzorec:
C₈H₄Br₂F₂O MW: 313.92156 g/mol |
MDL Number:
MFCD28738290 CAS číslo: 1807245-37-9 UN: 3261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...