Tisk…

4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide ≥95%

Dodavatel: Apollo Scientific

PC56686-250MG PC56686-1G
APOSPC56686-250MGEA 6220 CZK
APOSPC56686-250MG APOSPC56686-1G
4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide ≥95%
4-Bromo-2,5-bis(trifluoromethyl)benzyl bromide
Vzorec: C₉H₄Br₂F₆
MW: 385.92647 g/mol
MDL Number: MFCD28728007
CAS číslo: 1805513-09-0
UN: 3261

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR