Tisk…

2,4-Dibromo-6-fluorobenzyl bromide ≥95%

Dodavatel: Apollo Scientific

PC0746-250MG PC0746-1G
APOSPC0746-250MGEA 5390 CZK
APOSPC0746-250MG APOSPC0746-1G
2,4-Dibromo-6-fluorobenzyl bromide ≥95%
2,4-Dibromo-6-fluorobenzyl bromide
Vzorec: C₇H₄Br₃F
MDL Number: MFCD03094091
CAS číslo: 497181-23-4
UN: 3261

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR