Tisk…

3-Chloro-5-iodophenacyl bromide 95%

Dodavatel: Apollo Scientific

OR400227-250MG OR400227-1G
APOSOR400227-250MGEA 6420 CZK
APOSOR400227-250MG APOSOR400227-1G
3-Chloro-5-iodophenacyl bromide 95%
3-Chloro-5-iodophenacyl bromide
Vzorec: C₈H₅BrClIO
MDL Number: MFCD28040499
CAS číslo: 1823925-41-2
UN: 3261

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR