1-Bromo-2-chloro-4-fluoro-5-iodobenzene ≥95%
Dodavatel: Apollo Scientific
PC99064-250MG
PC99064-1G
PC99064-5G
APOSPC99064-250MGEA
6390
CZK
APOSPC99064-250MG
APOSPC99064-1G
APOSPC99064-5G
1-Bromo-2-chloro-4-fluoro-5-iodobenzene ≥95%
1-Bromo-2-chloro-4-fluoro-5-iodobenzene
Vzorec:
C₆H₂BrClFI |
MDL Number:
MFCD32903680 CAS číslo: 2460515-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...