Tisk…

2-Bromo-4-chloro-5-fluorophenol ≥95%

Dodavatel: Apollo Scientific

PC57127-250MG PC57127-1G
APOSPC57127-250MGEA 5200 CZK
APOSPC57127-250MG APOSPC57127-1G
2-Bromo-4-chloro-5-fluorophenol ≥95%
2-Bromo-4-chloro-5-fluorophenol
Vzorec: C₆H₃BrClFO
MW: 225.44282 g/mol
MDL Number: MFCD26895908
CAS číslo: 157165-23-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR