Tisk…

1-Bromo-2-chloro-4-(difluoromethoxy)benzene ≥95%

Dodavatel: Apollo Scientific

PC56983-250MG PC56983-1G PC56983-5G
APOSPC56983-250MGEA 910 CZK
APOSPC56983-250MG APOSPC56983-1G APOSPC56983-5G
1-Bromo-2-chloro-4-(difluoromethoxy)benzene ≥95%
1-Bromo-2-chloro-4-(difluoromethoxy)benzene
Vzorec: C₇H₄BrClF₂O
MDL Number: MFCD18390891
CAS číslo: 345226-22-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR