Tisk…

3,5-Dichloro-4-fluorobenzene-1,2-diamine ≥95%

Dodavatel: Apollo Scientific

PC57809-250MG PC57809-1G
APOSPC57809-250MGEA 5390 CZK
APOSPC57809-250MG APOSPC57809-1G
3,5-Dichloro-4-fluorobenzene-1,2-diamine ≥95%
3,5-Dichloro-4-fluorobenzene-1,2-diamine
Vzorec: C₆H₅Cl₂FN₂
MDL Number: MFCD29977177
CAS číslo: 1702399-91-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR