2-Bromo-4-chloro-5-fluorobenzyl alcohol ≥95%
Dodavatel: Apollo Scientific
PC57122-250MG
PC57122-1G
APOSPC57122-250MGEA
6850
CZK
APOSPC57122-250MG
APOSPC57122-1G
2-Bromo-4-chloro-5-fluorobenzyl alcohol ≥95%
2-Bromo-4-chloro-5-fluorobenzyl alcohol
Vzorec:
C₇H₅BrClFO |
MDL Number:
MFCD30184438 CAS číslo: 1067882-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...