4’-Bromo-2’,5’-bis(trifluoromethyl)acetophenone ≥95%
Dodavatel: Apollo Scientific
PC56694-250MG
PC56694-1G
APOSPC56694-250MGEA
7440
CZK
APOSPC56694-250MG
APOSPC56694-1G
4’-Bromo-2’,5’-bis(trifluoromethyl)acetophenone ≥95%
4’-Bromo-2’,5’-bis(trifluoromethyl)acetophenone
Vzorec:
C₁₀H₅BrF₆O MW: 335.04051 g/mol |
MDL Number:
MFCD28727780 CAS číslo: 1805971-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...