Tisk…

Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥95%

Dodavatel: Apollo Scientific

PC503028-250MG PC503028-1G
APOSPC503028-250MGEA 6220 CZK
APOSPC503028-250MG APOSPC503028-1G
Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate ≥95%
Methyl 5-chloro-2-fluoro-4-(trifluoromethyl)benzoate
Vzorec: C₉H₅ClF₄O₂
MW: 256.58141 g/mol
MDL Number: MFCD28752646
CAS číslo: 1805524-68-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR