Tisk…

4-Bromo-3,5-difluorobenzyl alcohol ≥95%

Dodavatel: Apollo Scientific

Synonyms: (4-Bromo-3,5-difluorophenyl)methanol

PC50177-250MG PC50177-1G
APOSPC50177-250MGEA 6850 CZK
APOSPC50177-250MG APOSPC50177-1G
4-Bromo-3,5-difluorobenzyl alcohol ≥95%
4-Bromo-3,5-difluorobenzyl alcohol
Vzorec: C₇H₅BrF₂O
MDL Number: MFCD22381218
CAS číslo: 1256276-36-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR