Tisk…

5-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine ≥95%

Dodavatel: Apollo Scientific

OR62037-1G OR62037-5G OR62037-10G
APOSOR62037-1GEA 2440 CZK
APOSOR62037-1G APOSOR62037-5G APOSOR62037-10G
5-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine ≥95%
5-Bromo-1-methyl-1H-pyrazolo[3,4-b]pyridine
Vzorec: C₇H₆BrN₃
MDL Number: MFCD16112604
CAS číslo: 887115-56-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR