Tisk…

2-Bromo-3-chloro-5-iodobenzyl alcohol ≥95%

Dodavatel: Apollo Scientific

OR61549-250MG OR61549-1G
APOSOR61549-250MGEA 6220 CZK
APOSOR61549-250MG APOSOR61549-1G
2-Bromo-3-chloro-5-iodobenzyl alcohol ≥95%
2-Bromo-3-chloro-5-iodobenzyl alcohol
Vzorec: C₇H₅BrClIO
CAS číslo: 2092867-48-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR