Tisk…

Methyl 2-ethoxy-4-nitro-benzoate ≥95%

Dodavatel: Apollo Scientific

OR52913-250MG OR52913-1G
APOSOR52913-250MGEA 4070 CZK
APOSOR52913-250MG APOSOR52913-1G
Methyl 2-ethoxy-4-nitro-benzoate ≥95%
Methyl 2-ethoxy-4-nitro-benzoate
Vzorec: C₁₀H₁₁NO₅
MW: 225.19804 g/mol
MDL Number: MFCD12025257
CAS číslo: 24091-87-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR