3,3,4,4-Tetramethylpyrrolidine-2,5-dione ≥98%
Dodavatel: Apollo Scientific
OR968325-250MG
OR968325-1G
OR968325-5G
APOSOR968325-250MGEA
2900
CZK
APOSOR968325-250MG
APOSOR968325-1G
APOSOR968325-5G
3,3,4,4-Tetramethylpyrrolidine-2,5-dione ≥98%
3,3,4,4-Tetramethylpyrrolidine-2,5-dione
Vzorec:
C₈H₁₃NO₂ |
MDL Number:
MFCD00047107 CAS číslo: 3566-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...