Tisk…

2-Amino-4-bromopyridin-3-ol hydrobromide ≥95%

Dodavatel: Apollo Scientific

OR914337-250MG OR914337-1G OR914337-5G
APOSOR914337-250MGEA 4440 CZK
APOSOR914337-250MG APOSOR914337-1G APOSOR914337-5G
2-Amino-4-bromopyridin-3-ol hydrobromide ≥95%
2-Amino-4-bromopyridin-3-ol hydrobromide
Vzorec: C₅H₆Br₂N₂O
MDL Number: MFCD09842810
CAS číslo: 114414-17-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR