Tisk…

5-Bromo-2-cyclopropylbenzoic acid ≥95%

Dodavatel: Apollo Scientific

OR70324-250MG OR70324-1G
APOSOR70324-250MGEA 6220 CZK
APOSOR70324-250MG APOSOR70324-1G
5-Bromo-2-cyclopropylbenzoic acid ≥95%
5-Bromo-2-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08126 g/mol
MDL Number: MFCD29067870
CAS číslo: 121146-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR