Tisk…

4-Bromo-2-cyclopropylbenzoic acid ≥95%

Dodavatel: Apollo Scientific

OR70322-250MG OR70322-1G
APOSOR70322-250MGEA 6220 CZK
APOSOR70322-250MG APOSOR70322-1G
4-Bromo-2-cyclopropylbenzoic acid ≥95%
4-Bromo-2-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08126 g/mol
MDL Number: MFCD31588710
CAS číslo: 1561780-76-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR