Tisk…

3-Bromo-4-cyclopropylbenzoic acid

Dodavatel: Apollo Scientific

OR70320-250MG OR70320-1G OR70320-5G
APOSOR70320-250MGEA 4470 CZK
APOSOR70320-250MG APOSOR70320-1G APOSOR70320-5G
3-Bromo-4-cyclopropylbenzoic acid
3-Bromo-4-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08 g/mol
MDL Number: MFCD11110859
CAS číslo: 1131622-50-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR