Tisk…

3-Bromo-2-cyclopropylbenzoic acid ≥95%

Dodavatel: Apollo Scientific

OR70319-250MG OR70319-1G
APOSOR70319-250MGEA 6220 CZK
APOSOR70319-250MG APOSOR70319-1G
3-Bromo-2-cyclopropylbenzoic acid ≥95%
3-Bromo-2-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08126 g/mol
MDL Number: MFCD32862617
CAS číslo: 2640734-74-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR