Tisk…

2-Bromo-4-cyclopropylbenzoic acid ≥97%

Dodavatel: Apollo Scientific

OR70317-250MG OR70317-1G
APOSOR70317-250MGEA 5790 CZK
APOSOR70317-250MG APOSOR70317-1G
2-Bromo-4-cyclopropylbenzoic acid ≥97%
2-Bromo-4-cyclopropylbenzoic acid
Vzorec: C₁₀H₉BrO₂
MW: 241,08126 g/mol
MDL Number: MFCD31715078
CAS číslo: 1237073-59-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR