Tisk…

2-Bromo-1-cyclopropyl-1H-imidazole ≥95%

Dodavatel: Apollo Scientific

OR302569-250MG OR302569-1G
APOSOR302569-250MGEA 7440 CZK
APOSOR302569-250MG APOSOR302569-1G
2-Bromo-1-cyclopropyl-1H-imidazole ≥95%
2-Bromo-1-cyclopropyl-1H-imidazole
Vzorec: C₆H₇BrN₂
MW: 187,03718 g/mol
MDL Number: MFCD19347862
CAS číslo: 1267001-46-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR