Tisk…

N-amino-6-chloropyridine-2-carboximidamide

Dodavatel: Apollo Scientific

OR71108-250MG OR71108-1G
APOSOR71108-250MGEA 827 CZK
APOSOR71108-250MG APOSOR71108-1G
N-amino-6-chloropyridine-2-carboximidamide
N-amino-6-chloropyridine-2-carboximidamide
Vzorec: C₆H₇ClN₄
MDL Number: MFCD34181144
CAS číslo: 2654087-89-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR