Tisk…

8-Bromo-4-cyanoquinoline-n-oxide ≥95%

Dodavatel: Apollo Scientific

OR70311-250MG OR70311-1G
APOSOR70311-250MGEA 6450 CZK
APOSOR70311-250MG APOSOR70311-1G
8-Bromo-4-cyanoquinoline-n-oxide ≥95%
8-Bromo-4-cyanoquinoline-n-oxide
Vzorec: C₁₀H₅BrN₂O
MDL Number: MFCD32862645
CAS číslo: 2640734-75-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR